EMAIL THIS PAGE TO A FRIEND
Neuroscience and behavioral physiology
Initial neurons of associative and callosal pathways of lateral suprasylvian area of the cat brain.
PMID 3561758
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
79311
Malvidin-3-galactoside chloride, analytical standard
C23H25ClO12