EMAIL THIS PAGE TO A FRIEND
Biomedizinische Technik. Biomedical engineering
[Monensin, an electrically charged carboxyl ionophor, and its neutral methylester as ion active components for Na+ selective disk electrodes].
PMID 3567283
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30552
Monensin methyl ester, Selectophore, ≥97.0% (TLC)
C37H64O11