EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Hygienic evaluation of working conditions during the conduct of aviation-chemical activities with ricid-II in agriculture].
PMID 3569972
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45814
Iprobenfos, PESTANAL®, analytical standard
C13H21O3PS