EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Experimental substantiation of an approximate safe level of p-chlorobenzaldehyde in the air of the work area].
PMID 3570017
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112216
4-Chlorobenzaldehyde, 97%
C7H5ClO