EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Determination of methylglyoxal as 2-methylquinoxaline by high-performance liquid chromatography and its application to biological samples.
PMID 3571379
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M80202
2-Methylquinoxaline, 97%
C9H8N2
W511609
2-Methylquinoxaline, ≥97%
C9H8N2