EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Biologically active coumarins from Enkleia siamensis.
PMID 3572421
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00193 Nordentatin, ≥95% (LC/MS-ELSD)
C19H20O4