EMAIL THIS PAGE TO A FRIEND
Meditsinskaia parazitologiia i parazitarnye bolezni
[Repellent effect of Mannich bases, derivatives of lactic acid esters].
PMID 3574247
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
316598
Isobutyl (R)-(+)-lactate, 97%
C7H14O3