EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Development of an enzyme-linked immunosorbent assay for the quantification of DDA (2,2-bis(p-chlorophenyl) acetic acid) in urine.
PMID 3580596
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35484
4,4′-DDA, PESTANAL®, analytical standard
C14H10Cl2O2
100870
Bis(4-chlorophenyl)acetic acid, 98%
C14H10Cl2O2