EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Migration of Irganox 1010 from ethylene-vinyl acetate films to foods and food-simulating liquids.
PMID 3583159
Abstract
In a series of experiments on the migration of the antioxidant Irganox 1010 from ethylene-vinyl acetate (EVA) films into food-simulating liquids and foods, the antioxidant was found to migrate rapidly from EVA film into n-heptane, 100% ethanol and corn oil. The rate of migration into these media was greater from EVA than from low-density polyethylene (LDPE) under comparable conditions. In contrast, little migration of Irganox 1010 was recorded on exposure of the EVA film to aqueous media, whereas migration from LDPE into such media was relatively high.