EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Effects of two organophosphorus insecticides on some biochemical constituents in the nervous system and liver of rabbits.
PMID 3584839
Abstract
Male Baladi rabbits were acutely and sub-chronically intoxicated with cyanofenphos and profenophos. The levels of cholesterol, triglycerides, B-lipoproteins and total proteins were determined in the serum, brain, spinal cord and sciatic nerve of rabbits. Moreover, the activities of alkaline phosphatase, acid phosphatase and glutamic-pyruvic transaminase were determined in the liver of the animals. The whole studies revealed that the biochemical constituents were highly affected by the tested insecticides. Also, the liver function suffered from adverse effects of the tested insecticides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33028
Cyanofenphos solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C15H14NO2PS