EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Metabolism of the hair dye component, nitro-p-phenylenediamine, in the rat.
PMID 3594688
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N21200
2-Nitro-1,4-phenylenediamine, 95%
C6H7N3O2