EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Quantification of pipecolic acid in plasma, and of urinary cystine in the presence of penicillamine-cystine disulfide, with the Beckman 6300 amino acid analyzer.
PMID 3594900
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1101
D-Penicillamine disulfide, 97%
C10H20N2O4S2