EMAIL THIS PAGE TO A FRIEND
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi
[Histological, histochemical and histoenzymatic changes at the level of the rat liver under the action of secbumetone].
PMID 3602727
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45658
Secbumeton, PESTANAL®, analytical standard
C10H19N5O