EMAIL THIS PAGE TO A FRIEND
Ukrainskii biokhimicheskii zhurnal (1978)
[Effect of thiamine and its derivatives on acetylcholinesterase activity of the blood and brain of albino mice].
PMID 3603737
Abstract
Thiamine, thiaminepyrophosphate and 4-methyl-5-beta-oxyethylthiazole are studied for their effect on the acetylcholinesterase activity in the brain, blood plasma and cells. The activity of acetylcholinesterase in blood cells is shown to be inhibited most of all by thiamine and thiazole. Acetylcholinesterase of the brain has been efficiently inhibited only by thiamine pyrophosphate.