EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The role of guanosine 5'-triphosphate in polypeptide chain elongation.
PMID 361078
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10106399001
GTP, Disodium salt
C10H16N5O14P3 · xNa+ · yH2O
G9002
Guanosine 5′-triphosphate tris salt, ≥93% (HPLC), powder
C10H16N5O14P3 · 3 C4H11NO3