EMAIL THIS PAGE TO A FRIEND
Die Nahrung
[The enzymatic degradation of DDT. 6. The fate of the DDT metabolites dichlorodiphenyldichloroethylene and dichlorodiphenylmonochloroethylene in the rat].
PMID 3614335
Abstract
DDE is hydrogenated in the rat organism. The distribution of the compound is described. DDMU is degradated to DBP as well as transformed to DDA and DDOH. Excretion and distribution are studied.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35484
4,4′-DDA, PESTANAL®, analytical standard
C14H10Cl2O2
100870
Bis(4-chlorophenyl)acetic acid, 98%
C14H10Cl2O2