EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
The active site of liver alcohol dehydrogenase mechanistic inferences from the binding and turnover of 2-, 3-, and 4-pyridylcarbinols.
PMID 3615419
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P66807
3-Pyridinemethanol, 98%
C6H7NO