EMAIL THIS PAGE TO A FRIEND
Zhongguo ji sheng chong xue yu ji sheng chong bing za zhi = Chinese journal of parasitology & parasitic diseases
[Effects of various anti-hookworm drugs on the cholinesterase activity of Necator americanus, Ancylostoma duodenale, Ancylostoma caninum and Nippostrongylus braziliensis].
PMID 3621504
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5650
Bephenium hydroxynaphthoate, analytical standard
C28H29NO4