EMAIL THIS PAGE TO A FRIEND
Giornale italiano di dermatologia e venereologia : organo ufficiale, Societa italiana di dermatologia e sifilografia
[Diflucortolone valerate and chlorquinaldol in topical eczema (in single or combination form)].
PMID 3623611
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130532
5,7-Dichloro-8-hydroxy-2-methylquinoline, 98%
C10H7Cl2NO