EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Separation of methyl-substituted benz[c]acridines by reversed-phase high-performance thin-layer chromatography.
PMID 3624385
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR158
Benz[c]acridine, BCR® certified Reference Material
C17H11N