EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Indazoles as indole bioisosteres: synthesis and evaluation of the tropanyl ester and amide of indazole-3-carboxylate as antagonists at the serotonin 5HT3 receptor.
PMID 3625701