EMAIL THIS PAGE TO A FRIEND
Wei sheng wu xue bao = Acta microbiologica Sinica
[Biosynthesis of nitrosopiperidine from isobutylamine and NaNo2 by some fungi].
PMID 3630140
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I14150
Isobutylamine, 99%
C4H11N