EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Influence of substrate concentration on the phase I and phase II metabolism of 4-methoxybiphenyl by rat isolated hepatocytes.
PMID 3630209
Abstract
1. The metabolism of 4-methoxybiphenyl to 4-hydroxybiphenyl and its sulphate and glucuronic acid conjugates has been studied in rat isolated hepatocytes at various concentrations of 4-methoxybiphenyl. 2. The proportions of metabolites produced remained constant at concentrations of 4-methoxybiphenyl less than 25-50 microM, but at higher concentrations the pattern of metabolism changed, there being proportionally more unconjugated 4-hydroxybiphenyl and its glucuronide, and less 4-hydroxybiphenyl sulphate. 3. The amount of 4-hydroxybiphenyl sulphate produced approached a plateau at the highest level of overall metabolism, whereas the amount of 4-hydroxybiphenyl glucuronide produced was linearly related to the amount of unconjugated 4-hydroxybiphenyl detected. 4. These data are consistent with the view that sulphation acts as a high-affinity, low-capacity pathway, whereas glucuronidation acts as a low-affinity, high-capacity pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
374776
4-Methoxybiphenyl, 97%
C13H12O