EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Location of conjugated diene position in an aliphatic chain by mass spectrometry of the 4-phenyl-1,2,4-triazoline-3,5-dione adduct.
PMID 3631517
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
280992
4-Phenyl-1,2,4-triazoline-3,5-dione, 97%
C8H5N3O2