EMAIL THIS PAGE TO A FRIEND
Soins. Psychiatrie
[Humoryl].
PMID 3634492
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3452
Toloxatone, ≥98% (HPLC), solid
C11H13NO3