EMAIL THIS PAGE TO A FRIEND
Bulletin des societes d'ophtalmologie de France
[Eye burn caused by sulfur monochloride].
PMID 3652349
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157759
Sulfur monochloride, 98%
Cl2S2