EMAIL THIS PAGE TO A FRIEND
Indian journal of experimental biology
Antioxidant protection mechanism of chick bone marrow: effects of toxicant induced stress.
PMID 3653935
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
449946
Neodymium(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Cl3Nd