EMAIL THIS PAGE TO A FRIEND
Journal of economic entomology
Use of acaricide-impregnated ear tags for controlling the brown ear tick (Acari: Ixodidae) in South Africa.
PMID 3655088
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33496
Flucythrinate, PESTANAL®, analytical standard, mixture of stereo isomers
C26H23F2NO4