EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
3-Mercaptopyruvate, 3-mercaptolactate and mercaptoacetate.
PMID 3657534
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
359394
Sodium mercaptopyruvate, ≥85%, technical grade
C3H3NaO3S