EMAIL THIS PAGE TO A FRIEND
Annales de biologie clinique
[Determination of blood iron: chromazurol B, ferrozine or the method selected by the SFBC (Société Française de Biologie Clinique)?].
PMID 3662140
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1782
Chromeazurol B
C23H14Cl2Na2O6