EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
A rapid test for chitinase activity that uses 4-methylumbelliferyl-N-acetyl-beta-D-glucosaminide.
PMID 3662513
Abstract
A total of 101 strains of bacteria from environmental and clinical sources, most of which were gram negative, were tested for chitobiase activity by using a filter paper spot test with 4-methylumbelliferyl-N-acetyl-beta-D-glucosaminide as the substrate. The results were compared with those obtained by a conventional plate method for chitinase activity by using colloidal chitin as the substrate. There was excellent agreement in the results for both methods. The filter paper spot test with 4-methylumbelliferyl-N-acetyl-beta-D-glucosaminide has the advantages of being rapid, simple to perform, and inexpensive. This method should be adaptable to a wider range of microorganisms, particularly those with unusual growth requirements.
Related Materials