EMAIL THIS PAGE TO A FRIEND
Beitrage zur gerichtlichen Medizin
[Enzyme histochemical studies of the myocardium following acethydrazine poisoning].
PMID 3663045
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8309
Acethydrazide, 90%
C2H6N2O