EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Determination of theophylline and its metabolites in plasma and urine by reversed-phase liquid chromatography using an amine modifier.
PMID 3667777
Abstract
A high-performance liquid chromatographic method for the determination of the concentrations of theophylline and its metabolites in plasma and urine samples is presented. The method uses the decylammonium ion, an N-alkylammonium modifier, and makes it possible to separate theophylline and its metabolites from other uric acid or xanthine derivatives, especially 1,7-dimethylxanthine, a metabolite of caffeine. The plasma and urine purification procedure is convenient, rapid and reproducible, and it can be used to determine low plasma and urine concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30692
Decylamine, ≥99.0% (GC)
C10H23N
D2404
Decylamine, 95%
C10H23N
442540
Decylamine, analytical standard
C10H23N