EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Method for determination of free and total glutathione and gamma-glutamylcysteine concentrations in human leukocytes and plasma.
PMID 3667817
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0903
γ-Glu-Cys, ≥80% (HPLC)
C8H14N2O5S