EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Reactions of oxobis(acetylacetonato)-vanadium (IV) with some purine and pyrimidine bases and nucleosides.
PMID 3668527
Abstract
A study of the reactions of purine and pyrimidine bases has been initiated with the V(IV) complex [V(O)(acac)2]. Two distinct modes of reaction have been recognized depending on the nature of the base. Simple adduct formation occurs when the base contains a sufficiently basic lone pair, but when bases contain acidic protons bound to nitrogen (inosine, thymine), cleavage of one acetylacetonate ligand occurs to give potentially polymeric species. This behavior is in contrast to that of the inert [Co(NO2)2(acac)2]-ion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
550787
Vanadyl acetylacetonate, 98%
C10H14O5V
94735
Vanadyl acetylacetonate, purum, ≥97.0% (RT)
C10H14O5V