EMAIL THIS PAGE TO A FRIEND
Photochemistry and photobiology
Chemistry of singlet oxygen--48. Isolation and structure of the primary product of photooxygenation of 3,5-di-t-butyl catechol.
PMID 3671512
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D45800
3,5-Di-tert-butylcatechol, 98%
C14H22O2