EMAIL THIS PAGE TO A FRIEND
Planta medica
Effects of cirsimaritin, a flavone isolated from Artemisia judaica, on isolated guinea-pig ileum.
PMID 3671552
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00174
Cirsimaritin, ≥90% (LC/MS-ELSD)
C17H14O6