EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
[Pharmacological studies of the central effects of new ethylguanidine, imidazolidine and imidazolidinone-2 derivatives].
PMID 3673624
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31534
2-Imidazolidone, PESTANAL®, analytical standard
C3H6N2O
I601
2-Imidazolidone, 96%
C3H6N2O