EMAIL THIS PAGE TO A FRIEND
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao
[Experimental study on SrCO3 preparation by double decomposition at room temperature].
PMID 3679169