EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Synthesis of the dipeptide derivatives of daunomycinone and adriamycinone].
PMID 3687444
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0120000 Daunorubicinone, European Pharmacopoeia (EP) Reference Standard
C21H18O8