EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Studies on the synthesis and neuromuscular-blocking activity of symmetrical bis-and poly-quaternary derivatives of quinuclidine and tropine].
PMID 3687459
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93550
Tropine, ≥97.0% (NT)
C8H15NO