EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Persistence studies with the herbicide, haloxyfop-methyl, in prairie field plots.
PMID 3690005
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45817
Haloxyfop, PESTANAL®, analytical standard
C15H11ClF3NO4