EMAIL THIS PAGE TO A FRIEND
International journal of peptide and protein research
Improved purities for Fmoc-amino acids from Fmoc-ONSu.
PMID 3692689
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46920
Fmoc N-hydroxysuccinimide ester, ≥98.0% (HPLC)
C19H15NO5
289507
Fmoc-OSu, 96%
C19H15NO5