EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Reversal of resistance by N-acetyltyramine or N-acetyl-2-phenylethylamine in doxorubicin-resistant leukemia P388 cells.
PMID 3693136
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00019
N-Acetyltyramine, ≥95% (LC/MS-ELSD)
C10H13NO2