EMAIL THIS PAGE TO A FRIEND
Journal of economic entomology
Efficacy of levamisole/famphur paste for control of cattle grubs (Diptera: Oestridae) and gastrointestinal worms (Nematoda: Trichostrongylidae).
PMID 3693663