EMAIL THIS PAGE TO A FRIEND
European journal of radiology
Ideal imaging in MR: contrast-enhancing pharmaceuticals. Evaluation of tempo carboxylic acid as nitroxide diagnostic agent.
PMID 3699037
Abstract
The meglumine salt of TCA, a piperidinyl nitroxide stable free radical derivative, was tested as an NMR contrast enhancer. This report of its use in animals indicates its usefulness in the enhancement of signal intensity of the kidney and bladder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
382000
4-Carboxy-TEMPO, free radical, 97%
C10H18NO3