EMAIL THIS PAGE TO A FRIEND
Fundamental and applied toxicology : official journal of the Society of Toxicology
"Potentiation of acute toxicity of 2-sec-butylphenyl N-methylcarbamate (BPMC) by fenthion in mice" by Miyaoka et al.
PMID 3699327
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45488
Fenobucarb, PESTANAL®, analytical standard
C12H17NO2