EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact urticaria to naphthylacetic acid.
PMID 3709145
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4002
2-Naphthaleneacetic acid, 99%
C12H10O2