EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Determination of tin compounds in the atmosphere by inversion volt-amperometry].
PMID 3710211
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36795
Sethoxydim, PESTANAL®, analytical standard
C17H29NO3S