EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Immunologic reactivity of patients with platinum poisoning].
PMID 3710256
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262587
Chloroplatinic acid solution, 8 wt. % in H2O
H2Cl6Pt